Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UQCRH Recombinant Rabbit Monoclonal Antibody (JE62-25)
GREENER_CHOICE

Catalog No. PIMA544807
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544807 Supplier Invitrogen™ Supplier No. MA544807
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

UQCRH Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow

Cytochrome b-c1 complex subunit 6, mitochondrial is a protein that in humans is encoded by the UQCRHgene. This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This protein may mediate formation of the complex between cytochromes c and c1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UQCRH
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE62-25
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene UQCRH
Gene Accession No. P07919, P99028, Q5M9I5
Gene Alias 2210416J04Rik; 2310021J10Rik; 2610041P16Rik; Complex III subunit 6; complex III subunit VIII; Cytochrome b-c1 complex subunit 6, mitochondrial; cytochrome c1 non-heme 11 kDa protein; cytochrome C1, nonheme 11 kDa protein; mitochondrial hinge protein; QCR6; ubiquinol cytochrome c reductase hinge protein; ubiquinol-cytochrome c reductase complex 11 kDa protein; ubiquinol-cytochrome c reductase hinge protein; ubiquinol-cytochrome c reductase, complex III subunit VIII; UQCR8; UQCRH
Gene Symbols UQCRH
Host Species Rabbit
Immunogen Synthetic peptide within human UQCRH aa 42-91/91.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 366448, 66576, 7388
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.