Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UQCRQ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565774 Supplier Invitrogen™ Supplier No. PA565774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IRESFFRVVPQ FVVFYLIYTW GTEEFERSKR KNPAAY Highest antigen sequence identity to the following orthologs - mouse 70%, rat 68%.

This gene encodes a ubiquinone-binding protein of low molecular mass. This protein is a small core-associated protein and a subunit of ubiquinol-cytochrome c reductase complex III, which is part of the mitochondrial respiratory chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UQCRQ
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UQCRQ
Gene Accession No. O14949
Gene Alias 1100001F06Rik; 1500040F11Rik; 5830407L17Rik; 9.5kDa; AA959903; c1502; Complex III subunit 8; complex III subunit VII; complex III subunit VIII; Cytochrome b-c1 complex subunit 8; low molecular mass ubiquinone-binding protein; low molecular mass ubiquinone-binding protein (9.5kD); low molecular mass ubiquinone-binding protein (Qpc-pending); MC3DN4; QCR8; QPC; QP-C; transcript under zygotic control; ubiquinol-cytochrome c reductase binding protein; ubiquinol-cytochrome c reductase complex 9.5 kDa protein; ubiquinol-cytochrome c reductase complex III subunit VII; ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C; ubiquinol-cytochrome c reductase, complex III subunit VII; ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5kDa; unknown; UQCR7; Uqcrb; UQCRQ
Gene Symbols UQCRQ
Host Species Rabbit
Immunogen Recombinant Human UQCRQ. Recombinant protein control fragment (Product #RP-106452).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27089
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.