Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uracil, 99.75%, MP Biomedicals™

Catalog No. ICN10320480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10320480 100 g
ICN10320425 25 g
ICN10320491 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10320480 Supplier MP Biomedicals, Inc Supplier No. 0210320480/DEL

Chemical Identifiers

CAS 66-22-8
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00006016
InChI Key ISAKRJDGNUQOIC-UHFFFAOYSA-N
Synonym uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x
PubChem CID 1174
ChEBI CHEBI:17568
IUPAC Name 1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES O=C1NC=CC(=O)N1

Specifications

Color White
Assay Percent Range 99.75%
Quantity 100 g
Formula Weight 112.09
Physical Form Crystalline Powder
Chemical Name or Material Uracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.