Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urapidil 98.0+%, TCI America™
SDP

Catalog No. U00855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00855G 5 g
U00851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00855G Supplier TCI America Supplier No. U00855G

Chemical Identifiers

CAS 34661-75-1
Molecular Formula C20H29N5O3
Molecular Weight (g/mol) 387.484
MDL Number MFCD00133908
InChI Key ICMGLRUYEQNHPF-UHFFFAOYSA-N
Synonym 6-[[3-[4-(2-Methoxyphenyl)-1-piperazinyl]propyl]amino]-1,3-dimethyluracil
PubChem CID 5639
IUPAC Name 6-[3-[4-(2-methoxyphenyl)piperazin-1-yl]propylamino]-1,3-dimethylpyrimidine-2,4-dione
SMILES CN1C(=CC(=O)N(C1=O)C)NCCCN2CCN(CC2)C3=CC=CC=C3OC

Specifications

Melting Point 159°C
Color White
Quantity 5 g
Formula Weight 387.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Urapidil
RTECSNumber YQ9862000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.