Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ URB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553787
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553787 Supplier Invitrogen™ Supplier No. PA553787
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VEHHKTCRSL GRSLWQQPSV GDILRLLDRD RMMQTILHFP QNRRLLPPED TQELIFKDKS RVDLDGLYDP CFLLQLFSEL TRPEFVVDCR KFLDSNALGL TVTALSSYDP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

URB1 gene ontology annotations related to this gene include maturation of 5.8S rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA); maturation of LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA).
TRUSTED_SUSTAINABILITY

Specifications

Antigen URB1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene URB1
Gene Accession No. O60287
Gene Alias 4921511H13Rik; 5730405K23Rik; AA670781; AI645590; C21orf108; KIAA0539; mKIAA0539; NOP254; NPA1; nucleolar pre-ribosomal-associated protein 1; nucleolar preribosomal-associated protein 1; nucleolar protein 254 kDa; Protein URB1; URB1; URB1 ribosome biogenesis 1 homolog; URB1 ribosome biogenesis 1 homolog (S. cerevisiae)
Gene Symbols URB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human URB1. Recombinant protein control fragment (Product #RP-91808).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9875
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.