Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ URB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531811
Description
Recommended positive controls: Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
URB2 gene ontology annotations related to this gene include ribosome biogenesis.
Specifications
| URB2 | |
| Polyclonal | |
| Unconjugated | |
| URB2 | |
| KIAA0133; NET10; NPA2; nucleolar preribosomal-associated protein 1; unhealthy ribosome biogenesis protein 2 homolog; URB2; URB2 ribosome biogenesis 2 homolog (S. cerevisiae) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9816 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q14146 | |
| URB2 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human URB2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction