Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, 98+%

Catalog No. AAA123600E
Click to view available options
Quantity:
10,000 g
2500 g
500 g

CAS: 57-13-6 | CH4N2O | 60.056 g/mol

Used as a protein denaturantUrea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a protein denaturantUrea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.

Solubility
Soluble in water, alcohol, methanol, glycerol, benzene and concentrated hydrochloric acid. Practically insoluble in chloroform and ether.

Notes
Stable at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 130°C to 135°C
Density 1.335
pH 9.2 to 9.5
Odor Ammonia-like
Quantity 2500 g
Beilstein 635724
Merck Index 14,9867
Solubility Information Soluble in water,alcohol,methanol,glycerol,benzene and concentrated hydrochloric acid. Practically insoluble in chloroform and ether.
Formula Weight 60.06
Percent Purity ≥98%
Chemical Name or Material Urea
Show More Show Less
EINECSNumber 200-315-5
RTECSNumber YR6250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.