Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, 99% Pure, Promega™

Catalog No. PRV3175 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-V3175 5 kg
PR-V3171 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR-V3175 Supplier Promega Corporation Supplier No. V3175
May include imposed supplier surcharges.
Add to Cart
Add to Cart

A protein denaturant qualified for use in polyacrylamide gels.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Color Colorless to White
Quantity 5 kg
Physical Form Crystalline Powder or Free-flowing Crystals
Chemical Name or Material Urea, For use as the denaturing component in polyacrylamide gels
Recommended Storage Store at 22° to 25°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.