Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, BiotechGrade, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18607652 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-652 12 kg Poly Pail
18-607-650 500 g Poly Bottle
18-607-651 2.5 kg Poly Pail
18-607-653 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-652 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. U102412KG
Add to Cart
Add to Cart

U1024, 57-13-6, CH4N2O

Spectrum™ Chemical Urea, BiotechGrade is an odorless and colorless solid that is an important nitrogen-containing substance found in mammal urine. In concentrations up to 10 M, urea is a powerful protein denaturant as it disrupts the noncovalent bonds in the proteins. This property can be exploited to increase the solubility of some proteins. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
IUPAC Name urea
SMILES NC(N)=O

Specifications

CAS Max % 1
Melting Point 132° to 134°C
Ignition Residue 0.001
Odor Odorless
Packaging Poly Pail
Quantity 12 kg
Percent Purity 99.0 to 100.5%
Grade Biotech
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.