Learn More
Urea (Colorless-to-White Crystals or Crystalline Powder/Mol. Biol.), Fisher BioReagents™
Shop All Fisherbrand ProductsDescription
Specifications
Specifications
| Absorbance | 0.05 max. (5M solution) at 280nm |
| Melting Point | 131°C |
| Color | White |
| pH | 7.5 to 9.5 |
| Assay Percent Range | ≥99 % |
| Packaging | Poly Drum |
| Linear Formula | NH2CONH2 |
| Quantity | 10 kg |
| Merck Index | 15, 10052 |
| Vapor Pressure | 1.25mmHg at 25°C |
| Show More |
Safety and Handling
| Recommended Storage | RT |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.