Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, MP Biomedicals

Catalog No. MP04821519 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
1 lb.
1 kg
25 lb.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04821519 1 lb.
MP04821518 50 g
MP04821532 25 lb.
MP04821528 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP04821519 Supplier MP Biomedicals, Inc Supplier No. 04821519
Add to Cart
Add to Cart

  • Dissolve urea in deionized water to the desired concentration
  • For every 10 ml of solution, add 1 g of Amberlite™ IRA-910
  • Stir for one hour at room temperature

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 130°C to 140°C
Color White
Boiling Point 135°C (decomposition)
Linear Formula CH4N2O
Quantity 1 lb.
Solubility Information One gram dissolves in 1 mL water, 10 mL 95% ethanol, 1 mL boiling 95% ethanol, 20 mL absolute ethanol, 6 mL methanol, 2 mL glycerol; Soluble in concentrated hydrochloric acid; almost insoluble in chloroform, ether.
Formula Weight 60.06
Percent Purity ≥99%
Grade Ultra Pure
Physical Form Prills
Chemical Name or Material UREA
Show More Show Less
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.