Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Urea, OmniPur™, Calbiochem™,

Catalog No. p-5584239
Click to view available options
Quantity:
500 g
5 kg

Ideal for protein unfolding/denaturation

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 132.8°C
Color White
pH 10.1 (25°C)
Quantity 500 g
Solubility Information Water: Soluble
Formula Weight 60.07
Concentration or Composition (by Analyte or Components) 0.9 to 1.1M
Physical Form Solid
Chemical Name or Material Urea
Grade Molecular biology grade
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.