Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Urea, USP, JP, 98-102%, Spectrum™ Chemical

Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Urea, USP, JP is used in topical dermatological products to promote rehydration of the skin. Urea preparations are also used for non-surgical removal of damaged or dead nails. All Spectrum Chemical USP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 1 |
Ignition Residue | 0.001 |
Packaging | Poly Pail |
Quantity | 2.5 kg |
Percent Purity | 98 to 102% |
Grade | Bio Certified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction