Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ureaplasma parvum Monoclonal Antibody (C46G)
GREENER_CHOICE

Catalog No. PIMA517010
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517010 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517010 Supplier Invitrogen™ Supplier No. MA517010
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17010 has been used in ELISA and IF applications and shows reactivity with the multi-banded antigen (MB) of several Ureaplasma serotypes. MA5-07010 does not cross react with Mycoplasma hominis and Mycoplasma genitallium. MA5-17010 reacts with serovars 1,2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12 & 14.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ureaplasma parvum
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C46G
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias U. parvum
Host Species Mouse
Immunogen Reactive with the multi-banded antigen (MB) of several Ureaplasma serotypes.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.