Learn More
Invitrogen™ Ureaplasma parvum Monoclonal Antibody (G9C)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA517011
Description
MA5-17011 has been used in ELISA and IF applications and shows reactivity with the multi-banded antigen (MB) of several Ureaplasma serotypes. MA5-07011 does not cross react with Mycoplasma hominis and Mycoplasma genitallium. MA5-17011 reacts with serovars: 1, 3, 6 & 14 (+ 7 & 11 weakly).

Specifications
Ureaplasma parvum | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA, Western Blot, Immunocytochemistry | |
G9C | |
Unconjugated | |
U. parvum | |
Reactive with the multi-banded antigen (MB) of several Ureaplasma serotypes. | |
1 mL | |
Primary | |
4°C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.