Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urease Polyclonal Antibody, HRP, Invitrogen™

Catalog No. PIPA128969
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PIPA128969 1 mg
1 options

Catalog No. PIPA128969

Supplier: Thermo Scientific PA128969

Rabbit Polyclonal Antibody

PA1-28969 detects Urease from plant samples.

Urease Polyclonal antibody specifically detects Urease in Plant samples. It is validated for Dot Blot, ELISA, Immunomicroscopy, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Urease
Applications Dot Blot, ELISA, Immunomicroscopy, Western Blot
Classification Polyclonal
Concentration 10 mg/mL
Conjugate HRP
Formulation PBS with 10mg/mL BSA, 50% glycerol and 0.01% gentamicin sulfate; pH 7.2
Gene Alias AT1G67550; F12B7.10; F12B7_10; URE; Urea amidohydrolase; urease
Host Species Rabbit
Immunogen Full length native protein: Urease (Jack Bean)
Purification Method Ion-exchange chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.