Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ URG4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544534
Description
Peptide sequence: AILHAFLRLE KTGHMPNYQF VYQNLHDVSV PGPRPRDKRQ LLDPPGDLSR Sequence homology: Human: 100%; Mouse: 86%; Rat: 86%.
The specific function of this protein remains unknown. URG4 is upregulated in the presence of hepatitis B virus (HBV)-encoded X antigen (HBxAg) and may contribute to the development of hepatocellular carcinoma by promoting hepatocellular growth and survival (Tufan et al., 2002 ).
Specifications
URG4 | |
Polyclonal | |
Unconjugated | |
URGCP | |
2010005J08Rik; 9130001I21Rik; AW060359; DKFZp666G166; DKFZp686O0457; HBV X protein up-regulated gene 4 protein; HBV X protein up-regulated gene 4 protein homolog; HBxAg up-regulated gene 4 protein; HBxAg up-regulated gene 4 protein homolog; KIAA1507; mKIAA1507; protein URG4; RGD1564681; up-regulated gene 4; upregulator of cell proliferation; up-regulator of cell proliferation; URG4; Urgcp | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55665 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TCY9 | |
URGCP | |
Synthetic peptide directed towards the middle region of human URG4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction