Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ URG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556160
Description
Immunogen sequence: HVRLERRSRL VVLSTVGVPG TGKSTLLNTM FGLRFATGKS CGPRGAFMQL ITVAEGFSQD LGCDHILVID SGGLIGGALT SAGDRFELEA SLATLLMGLS NVTVISLAET KD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
The specific function of this protein remains unknown. URG4 is upregulated in the presence of hepatitis B virus (HBV)-encoded X antigen (HBxAg) and may contribute to the development of hepatocellular carcinoma by promoting hepatocellular growth and survival (Tufan et al., 2002 ).
Specifications
| URG4 | |
| Polyclonal | |
| Unconjugated | |
| URGCP | |
| 2010005J08Rik; 9130001I21Rik; AW060359; DKFZp666G166; DKFZp686O0457; HBV X protein up-regulated gene 4 protein; HBV X protein up-regulated gene 4 protein homolog; HBxAg up-regulated gene 4 protein; HBxAg up-regulated gene 4 protein homolog; KIAA1507; mKIAA1507; protein URG4; RGD1564681; up-regulated gene 4; upregulator of cell proliferation; up-regulator of cell proliferation; URG4; Urgcp | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55665 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TCY9 | |
| URGCP | |
| Recombinant protein corresponding to Human URG4. Recombinant protein control fragment (Product #RP-92686). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction