Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

URG4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984557
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4557 Supplier Bioss Supplier No. BS2171R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The specific function of this protein remains unknown. URG4 is upregulated in the presence of hepatitis B virus (HBV)-encoded X antigen (HBxAg) and may contribute to the development of hepatocellular carcinoma by promoting hepatocellular growth and survival (Tufan et al., 2002 ).

Specifications

Antigen URG4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene URGCP
Gene Accession No. Q8TCY9
Gene Alias 2010005J08Rik; 9130001I21Rik; AW060359; DKFZp666G166; DKFZp686O0457; HBV X protein up-regulated gene 4 protein; HBV X protein up-regulated gene 4 protein homolog; HBxAg up-regulated gene 4 protein; HBxAg up-regulated gene 4 protein homolog; KIAA1507; mKIAA1507; protein URG4; RGD1564681; up-regulated gene 4; upregulator of cell proliferation; Up-regulator of cell proliferation; Urg4; Urgcp
Gene Symbols URGCP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human URG4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55665
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.