Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Uridine-5'-diphosphate disodium salt, 98%

Click to view available options
Quantity:
100mg
1g
27821-45-0
C9H12N2Na2O12P2
448.124
MFCD00149253
HTFLYOJAWHVZRW-WFIJOQBCSA-N
5'-udp,2na
131676107
disodium;(2R,3R,4S,5R)-2-(2,4-dioxopyrimidin-1-yl)-5-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolane-3,4-diolate
C1=CN(C(=O)NC1=O)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)O)[O-])[O-].[Na+].[Na+]
27821-45-0
C9H12N2Na2O12P2
448.124
MFCD00149253
HTFLYOJAWHVZRW-WFIJOQBCSA-N
5'-udp,2na
131676107
disodium;(2R,3R,4S,5R)-2-(2,4-dioxopyrimidin-1-yl)-5-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolane-3,4-diolate
C1=CN(C(=O)NC1=O)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)O)[O-])[O-].[Na+].[Na+]
Applications
Uridine-5′-diphosphate disodium salt is used in studies on nucleic acid (RNA) biosynthesis and cell signaling. UDP is a nucleotide that upon phosphorylation to UTP becomes a substrate for enzymes such as RNA polymerase(s) and GTPases. These enzymes are involved in a wide range of applications from the synthesis of RNA to the regulation of G-coupled Protein Receptors (GPCR) and cell signaling molecules such as Rho-signaling via Guanine Nucleotide Exchange Factors (GEF).

Solubility
Slightly soluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
Sensitivity Moisture Sensitive
Quantity 100mg
Color White
Beilstein 8817400
Formula Weight 448.13
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Uridine-5'-diphosphate disodium salt

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C

Recommended Storage : −30° to −10°C