Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uridine, ∽99%, MP Biomedicals™

Catalog No. ICN10321625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10321625 25 g
ICN10321605 5 g
ICN10321610 10 g
ICN10321680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10321625 Supplier MP Biomedicals, Inc Supplier No. 0210321625
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58-96-8
Molecular Formula C9H12N2O6
Molecular Weight (g/mol) 244.203
InChI Key DRTQHJPVMGBUCF-DHRBYNEYSA-N
Synonym uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd
PubChem CID 89495556
IUPAC Name 1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O

Specifications

Color White
Assay Percent Range ∽99%
Quantity 25 g
Formula Weight 244.2
Physical Form Powder
Chemical Name or Material Uridine
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.