Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uridine Phosphorylase 1/UPP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP155354
Description
Uridine Phosphorylase 1/UPP1 Polyclonal specifically detects Uridine Phosphorylase 1/UPP1 in Human samples. It is validated for Western Blot.Specifications
Uridine Phosphorylase 1/UPP1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
EC 2.4.2.3, UDRPASE, UPASE, UPP, UPUPase 1, UrdPase 1, uridine phosphorylase, uridine phosphorylase 1 | |
Rabbit | |
34 kDa | |
100 μL | |
Primary | |
Porcine: 75%; Guinea pig: 75%. | |
Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q16831 | |
UPP1 | |
Synthetic peptide directed towards the N terminal of human UPP1(NP_003355). Peptide sequence AATGANAEKAESHNDCPVRLLNPNIAKMKEDILYHFNLTTSRHNFPALFG. | |
Affinity purified | |
RUO | |
7378 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction