Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ URM1 Monoclonal Antibody (182CT27.2.2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537497
Description
URM1 Monoclonal Antibody for Western Blot
Following the discovery of protein modification by the small, highly conserved ubiquitin polypeptide, a number of distinct ubiquitin-like proteins (Ubls) have been found to function as protein modifiers as well. These Ubls, which include SUMO, ISG15, Nedd8, and Atg8, function as critical regulators of many cellular processes, including transcription, DNA repair, signal transduction, autophagy, and cell-cycle control. A growing body of data also implicates the dysregulation of Ubl-substrate modification and mutations in the Ubl-conjugation machinery in the etiology and progression of a number of human diseases. URM1 is a ubiquitin-like modifier protein.
Specifications
URM1 | |
Monoclonal | |
Unconjugated | |
Urm1 | |
2900073H19Rik; C9orf74; MGC2668; RGD1306599; RP11-339B21.4; ubiquitin related modifier 1; ubiquitin related modifier 1 homolog; ubiquitin related modifier 1 homolog (S. cerevisiae); ubiquitin-related modifier 1; ubiquitin-related modifier 1 homolog; ubiquitin-related modifier URM1; URM1; YIL008W | |
Mouse | |
Euglobulin Precipitation | |
RUO | |
81605 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
182CT27.2.2 | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9BTM9 | |
Urm1 | |
URM1 recombinant protein. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction