Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ URM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565807
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565807 Supplier Invitrogen™ Supplier No. PA565807
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVEVEFGGGAE LLFDGIKKHR VTLPGQEEPW DIRNLLIWIK KNLLKERPEL FIQGDSVRPG ILVLINDADW ELLGELDYQL QDQDSVLFIS TLHG Highest antigen sequence identity to the following orthologs - mouse 93%, rat 94%.

Following the discovery of protein modification by the small, highly conserved ubiquitin polypeptide, a number of distinct ubiquitin-like proteins (Ubls) have been found to function as protein modifiers as well. These Ubls, which include SUMO, ISG15, Nedd8, and Atg8, function as critical regulators of many cellular processes, including transcription, DNA repair, signal transduction, autophagy, and cell-cycle control. A growing body of data also implicates the dysregulation of Ubl-substrate modification and mutations in the Ubl-conjugation machinery in the etiology and progression of a number of human diseases. URM1 is a ubiquitin-like modifier protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen URM1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Urm1
Gene Accession No. Q9BTM9
Gene Alias 2900073H19Rik; C9orf74; MGC2668; RGD1306599; RP11-339B21.4; ubiquitin related modifier 1; ubiquitin related modifier 1 homolog; ubiquitin related modifier 1 homolog (S. cerevisiae); ubiquitin-related modifier 1; ubiquitin-related modifier 1 homolog; ubiquitin-related modifier URM1; URM1; YIL008W
Gene Symbols Urm1
Host Species Rabbit
Immunogen Recombinant Human URM1. Recombinant protein control fragment (Product #RP-103586).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81605
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.