Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UROC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584829
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584829 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584829 Supplier Invitrogen™ Supplier No. PA584829
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TDSPFRETSN IYDGSAFCAD MAVQNFVGDA CRGATWVALH NGGGVGWGEV INGGFGLVLD GTPEAEGRAR LMLSWDVSNG V Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 95%.

This gene encodes an enzyme involved in histidine catabolism, metabolizing urocanic acid to formiminoglutamic acid. The gene product is known to protect the skin from ultra violet rays and is contained in human sweat. Deficiency of this gene product in the liver is an apparent cause of mental retardation. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UROC1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UROC1
Gene Accession No. Q96N76
Gene Alias AI265605; BC022133; HMFN0320; Imidazolonepropionate hydrolase; UROC1; Urocanase; urocanase 1; urocanase domain containing 1; urocanate hydratase; urocanate hydratase 1; UROCD
Gene Symbols UROC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UROC1. Recombinant protein control fragment (Product #RP-108246).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131669
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.