Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UROD Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5143018
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
This gene encodes the fifth enzyme of the heme biosynthetic pathway. This enzyme is responsible for catalyzing the conversion of uroporphyrinogen to coproporphyrinogen through the removal of four carboxymethyl side chains. Mutations and deficiency in this enzyme are known to cause familial porphyria cutanea tarda and hepatoerythropoetic porphyria.
Specifications
| UROD | |
| Polyclonal | |
| Unconjugated | |
| Urod | |
| AI323803; PCT; porphyrinogen carboxylase; porphyrinogen carboxy-lyase; UPD; URO D; UROD; Uro-d; Uroporphyrinogen decarboxylase; uroporphyrinogen III decarboxylase | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 7389 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P06132 | |
| Urod | |
| Peptide with sequence KARECVGKTVTLQGN (aa 290-304). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction