Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UROD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555939
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555939 Supplier Invitrogen™ Supplier No. PA555939
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPGKGPSFPE PLREEQALDR LRDPEVVASE LGYVFQAITL TRQRLAGRVP LIGFAGAPWT LMTYMVEGGG SSTMAQAKRW LYQRPQASHQ LLRILTD Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.

This gene encodes the fifth enzyme of the heme biosynthetic pathway. This enzyme is responsible for catalyzing the conversion of uroporphyrinogen to coproporphyrinogen through the removal of four carboxymethyl side chains. Mutations and deficiency in this enzyme are known to cause familial porphyria cutanea tarda and hepatoerythropoetic porphyria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UROD
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Urod
Gene Accession No. P06132, P32362, P70697
Gene Alias AI323803; PCT; porphyrinogen carboxylase; porphyrinogen carboxy-lyase; UPD; URO D; UROD; Uro-d; Uroporphyrinogen decarboxylase; uroporphyrinogen III decarboxylase
Gene Symbols Urod
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UROD. Recombinant protein control fragment (Product #RP-94173).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22275, 29421, 7389
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.