Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Urokinase Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144658
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144658 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144658 Supplier Invitrogen™ Supplier No. PA5144658
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: PC-3M cell lysate, MCF-7 cell lysate, LANCAP cell lysate, rat kidney tissue lysate, rat stomach tissue lysate, PC-3M. Predicted band size: 49 kDa Subcellular Location: Secreted.

This gene encodes a serine protease involved in degradation of the extracellular matrix and possibly tumor cell migration and proliferation. A specific polymorphism in this gene may be associated with late-onset Alzheimer's disease and also with decreased affinity for fibrin-binding. This protein converts plasminogen to plasmin by specific cleavage of an Arg-Val bond in plasminogen. Plasmin in turn cleaves this protein at a Lys-Ile bond to form a two-chain derivative in which a single disulfide bond connects the amino-terminal A-chain to the catalytically active, carboxy-terminal B-chain. This two-chain derivative is also called HMW-uPA (high molecular weight uPA). HMW-uPA can be further processed into LMW-uPA (low molecular weight uPA) by cleavage of chain A into a short chain A (A1) and an amino-terminal fragment. LMW-uPA is proteolytically active but does not bind to the uPA receptor. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Urokinase
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PLAU
Gene Accession No. P00749, P29598
Gene Alias antagonist of uPA; ATF; BDPLT5; CD87; HGNC:9052; plasminogen activator; plasminogen activator, urinary; plasminogen activator, urokinase; Plau; QPD; uPA; u-PA; UPAM; UPAR; U-plasminogen activator; Urinary plasminogen activator, urokinase; URK; URKR; Urokinase-type plasminogen activator; Urokinase-type plasminogen activator chain B; Urokinase-type plasminogen activator long chain A; Urokinase-type plasminogen activator short chain A
Gene Symbols PLAU
Host Species Rabbit
Immunogen Recombinant protein within Human Urokinase 10-200.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25619, 5328
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.