Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Urolithin A
GREENER_CHOICESDP

Catalog No. 676250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-625-0 50 mg
67-621-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-625-0 Supplier Tocris Bioscience™ Supplier No. 6762/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

PI 3-K/Akt/mTOR signaling pathway inhibitor; blocks phosphorylation of Akt and p70S6K in pancreatic ductal adenocarcinoma (PDAC)

Urolithin A is a PI 3-K/Akt/mTOR signaling pathway inhibitor; blocks phosphorylation of Akt and p70S6K in pancreatic ductal adenocarcinoma (PDAC). Inhibits proliferation and migration and promotes apoptosis of PDAC cell lines in vitro (IC50 values are 10.23 - 62.28 μM). Suppresses tumor growth and improves survival in mice bearing pancreatic tumor xenografts. Also blocks amyloid-β fibrillation in vitro.

Specifications

Chemical Name or Material 3,8-Dihydroxy-6H-dibenzo[b,d]pyran-6-one
CAS 1143-70-0
Quantity 50 mg
Molecular Formula C13H8O4
Formula Weight 228.2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.