Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uromodulin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | Uromodulin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Uromodulin Polyclonal specifically detects Uromodulin in Human samples. It is validated for Western Blot.Specifications
Uromodulin | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication | |
ADMCKD2, FJHN, HNFJ, HNFJ1, MCKD2, Tamm-Horsfall glycoprotein, Tamm-Horsfall urinary glycoprotein, THGP, THP, uromodulin, uromodulin (uromucoid, Tamm-Horsfall glycoprotein), uromucoid | |
UMOD | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
P07911 | |
7369 | |
Synthetic peptides corresponding to UMOD(uromodulin) The peptide sequence was selected from the middle region of UMOD. Peptide sequence MTNCYATPSSNATDPLKYFIIQDRCPHTRDSTIQVVENGESSQGRFSVQM. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title