Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Uromodulin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537902
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537902 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537902 Supplier Invitrogen™ Supplier No. PA537902
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

This gene encodes uromodulin, the most abundant protein in normal urine. Its excretion in urine follows proteolytic cleavage of the ectodomain of its glycosyl phosphatidylinosital-anchored counterpart that is situated on the luminal cell surface of the loop of Henle. Uromodulin may act as a constitutive inhibitor of calcium crystallization in renal fluids. Excretion of uromodulin in urine may provide defense against urinary tract infections caused by uropathogenic bacteria. Defects in this gene are associated with the autosomal dominant renal disorders medullary cystic kidney disease-2 (MCKD2) and familial juvenile hyperuricemic nephropathy (FJHN). These disorders are characterized by juvenile onset of hyperuricemia, gout, and progressive renal failure. While several transcript variants may exist for this gene, the full-length natures of only two have been described to date. These two represent the major variants of this gene and encode the same isoform.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Uromodulin
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Umod
Gene Accession No. P07911
Gene Alias ADMCKD2; FJHN; HNFJ; HNFJ1; MCKD2; Tamm-Horsfall urinary glycoprotein; THGP; THP; UMOD; Urehd1; urehr4; Urmodulin (Tamm-Horsfall protein); uromodulin; Uromodulin, secreted form; uromucoid; uromucoid uromucoid
Gene Symbols Umod
Host Species Goat
Immunogen Peptide with sequence DRCPHTRDSTIQ, from the internal region of UMOD (aa525-536).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7369
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.