Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uroplakin-3 Monoclonal Antibody (ABM129), Abbomax

Catalog No. 502204201
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4201 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4201 Supplier Abbomax Inc Supplier No. 603950
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: PC-3 cell lysate Cellular location: Plasma membrane.

Uroplakin-3 Monoclonal antibody specifically detects Uroplakin-3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Uroplakin-3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone ABM129
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene UPK3B
Gene Accession No. O75631, Q9BT76
Gene Alias LOC105375355; p35; RGD1309615; UP3a; UP3b; UP3KA; UP3KB; UPIII; UPIIIA; UPIIIb; UPK3; Upk3a; UPK3A/B; Upk3b; uroplakin 3; uroplakin 3A; uroplakin 3B; uroplakin III; uroplakin IIIA; uroplakin IIIb; Uroplakin-3a; uroplakin-3b; urothelial differentiation-dependent marker
Host Species Mouse
Immunogen A synthetic peptide from C-term of human uroplakin-3 protein
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105375355, 7380
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.