Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uroplakin Ia Antibody (UPK1A/2921), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB011431 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB011431 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB011431 Supplier Novus Biologicals Supplier No. NBP279868AF700
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Uroplakin Ia Monoclonal antibody specifically detects Uroplakin Ia in Human, Canine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen Uroplakin Ia
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone UPK1A/2921
Conjugate Alexa Fluor 700
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA
Gene Symbols UPK1A
Host Species Mouse
Immunogen Recombinant fragment (around aa 114-173) of human Uroplakin Ia (UPK1A) protein (exact sequence is proprietary) (Uniprot: O00322)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.