Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uroplakin Ia Antibody (UPK1A/2922), Alexa Fluor™ 532, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP279869AF532

Catalog No. NB011450


Only null left
Add to Cart

Description

Description

Uroplakin Ia Monoclonal specifically detects Uroplakin Ia in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Uroplakin Ia
Monoclonal
Alexa Fluor 532
50mM Sodium Borate with 0.05% Sodium Azide
UPK1A
Recombinant fragment (around aa 114-173) of human Uroplakin Ia (UPK1A) protein (exact sequence is proprietary) (Uniprot: O00322)
0.1 mL
Primary
Store at 4°C in the dark.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
UPK1A/2922
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA
Mouse
Protein A or G purified
RUO
Human
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.