Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uroplakin Ia Mouse anti-Human, Clone: UPK1A/2923, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Uroplakin Ia |
---|---|
Clone | UPK1A/2923 |
Concentration | 0.2 mg/ml |
Dilution | ELISA, Protein Array |
Applications | ELISA, Peptide Array |
Description
Uroplakin Ia Monoclonal specifically detects Uroplakin Ia in Human samples. It is validated for ELISA, Protein Array.Specifications
Uroplakin Ia | |
0.2 mg/ml | |
ELISA, Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA | |
Recombinant fragment (around aa 114-173) of human Uroplakin Ia protein (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
UPK1A/2923 | |
ELISA, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
11045 | |
IgG2b κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title