Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uroplakin Ia, Mouse anti-Human, Clone: UPK1A/2925, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP27973520UG
Description
Uroplakin Ia Monoclonal specifically detects Uroplakin Ia in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Uroplakin Ia | |
Monoclonal | |
0.2 mg/mL | |
ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA | |
Mouse | |
Protein A or G purified | |
RUO | |
11045 | |
Store at 4°C. | |
IgG1 κ |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
UPK1A/2925 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
UPK1A | |
Recombinant fragment (around aa 114-173) of human Uroplakin Ia protein (exact sequence is proprietary) | |
20 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction