Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uroplakin Ia Antibody (UPK1A/2922), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279869G
Description
Uroplakin Ia Monoclonal specifically detects Uroplakin Ia in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Uroplakin Ia | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
UPK1A | |
Recombinant fragment (around aa 114-173) of human Uroplakin Ia (UPK1A) protein (exact sequence is proprietary) (Uniprot: O00322) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
UPK1A/2922 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction