Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uroplakin Ib Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281768F
This item is not returnable.
View return policy
Description
Uroplakin Ib Polyclonal antibody specifically detects Uroplakin Ib in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Uroplakin Ib | |
Polyclonal | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
FITC | |
tetraspanin-20, Tspan-20, TSPAN20tetraspan, UP1b, UPIb, UPK1, uroplakin 1B, Uroplakin Ib, uroplakin-1b | |
Uroplakin Ib antibody was raised against a 17 amino acid synthetic peptide near the center of human Uroplakin Ib. The immunogen is located within amino acids 110 - 160 of Uroplakin Ib. Amino Acid Squence: KQMLERYQNNSPPNNDD | |
0.1 mL | |
Signal Transduction | |
7348 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction