Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uroplakin Ib, Mouse anti-Human, Clone: UPK1B/3102, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279800100UG
Description
Uroplakin Ib Monoclonal specifically detects Uroplakin Ib in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Uroplakin Ib | |
Monoclonal | |
0.2 mg/mL | |
ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
tetraspanin-20, Tspan-20, TSPAN20tetraspan, UP1b, UPIb, UPK1, uroplakin 1B, Uroplakin Ib, uroplakin-1b | |
Mouse | |
Protein A or G purified | |
RUO | |
7348 | |
Store at 4°C. | |
IgG1 κ |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
UPK1B/3102 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
UPK1B | |
Recombinant fragment (around aa 109-229) of human Uroplakin Ib protein (exact sequence is proprietary) | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction