Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uroplakin IIIB Antibody (UPK3B/8550R), Novus Biologicals™

Rabbit Monoclonal Antibody
$247.50 - $474.50
Specifications
Antigen | Uroplakin IIIB |
---|---|
Clone | UPK3B/8550R |
Concentration | 0.2 mg/ml |
Dilution | ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL |
Applications | ELISA, Immunohistochemistry (Paraffin) |
Description
Uroplakin IIIB Monoclonal antibody specifically detects Uroplakin IIIB in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
Uroplakin IIIB | |
0.2 mg/ml | |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
10mM PBS with 0.05% BSA | |
105375355 | |
IgG κ | |
Protein A or G purified |
UPK3B/8550R | |
ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
FLJ32198, MGC10902, p35, UP3b, UPIIIB, UPIIIbP35, uroplakin 3B, Uroplakin IIIb, uroplakin-3b | |
Recombinant fragment (around aa 100-300) of human Uroplakin IIIB protein (exact sequence is proprietary) | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title