Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ US12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144429 Supplier Invitrogen™ Supplier No. PA5144429
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

US12 Polyclonal Antibody for Western Blot, ELISA

Plays a role in the inhibition of host immune response. Binds specifically to transporters associated with antigen processing (TAP), thereby blocking peptide-binding and translocation by TAP as well as subsequent loading of peptides onto MHC class I molecules. Empty MHC I molecules are retained in the endoplasmic reticulum and ultimately directed to proteasomal degradation. In consequence, infected cells are masked for immune recognition by cytotoxic T-lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen US12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.249 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene US12
Gene Accession No. P03170
Gene Alias ICP47 protein; US12
Gene Symbols US12
Host Species Rabbit
Immunogen Recombinant Human herpesvirus 1 ICP47 protein (US12) (1-88aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2703441
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.