Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USAG1/SOSTDC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | USAG1/SOSTDC1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
USAG1/SOSTDC1 Polyclonal specifically detects USAG1/SOSTDC1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
USAG1/SOSTDC1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
cystine-knot containing secreted protein, DKFZp564D206, Ectodermal BMP inhibitor, ECTODIN, sclerostin domain containing 1, sclerostin domain-containing protein 1, USAG-1, USAG1uterine sensitization-associated protein-1, Uterine sensitization-associated gene 1 protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 115-206 of human USAG1/SOSTDC1 (NP_056279.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS with 50% glycerol, pH7.3. | |
25928 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title