Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USE1 Polyclonal Antibody
USE1 Polyclonal Antibody
Supplier: Thermo Scientific PA541595
Description
USE1 Polyclonal Antibody for Western Blot, IHC

Specifications
USE1 | |
Polyclonal | |
Unconjugated | |
Use1 | |
2010315L10Rik; 5730403H22Rik; AV002165; D12; Ed2; embryonic development factor 2; fj43b06; hypothetical protein MGC56402; MDS032; P31; Protein D12; protein p31; putative MAPK activating protein PM26; putative MAPK-activating protein PM26; Q-snare; RGD1306660; SLT1; SNARE-like tail-anchored protein 1 homolog; unconventional SNARE in the ER 1; unconventional SNARE in the ER 1 homolog; unconventional SNARE in the ER 1 homolog (S. cerevisiae); USE1; Use1l; USE1-like protein; Vesicle transport protein USE1; wu:fj43b06; zgc:56402 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55850 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NZ43 | |
Use1 | |
Synthetic peptide directed towards the N-terminal of human MDS032 (aa 7-56). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction