Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589024 Supplier Thermo Scientific Supplier No. PA589024
Add to Cart
Add to Cart

USE1 Polyclonal Antibody

USE1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen USE1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Use1
Gene Accession No. Q9CQ56
Gene Alias 2010315L10Rik; 5730403H22Rik; AV002165; D12; Ed2; embryonic development factor 2; fj43b06; hypothetical protein MGC56402; MDS032; P31; Protein D12; protein p31; putative MAPK activating protein PM26; putative MAPK-activating protein PM26; Q-snare; RGD1306660; SLT1; SNARE-like tail-anchored protein 1 homolog; unconventional SNARE in the ER 1; unconventional SNARE in the ER 1 homolog; unconventional SNARE in the ER 1 homolog (S. cerevisiae); USE1; Use1l; USE1-like protein; Vesicle transport protein USE1; wu:fj43b06; zgc:56402
Gene Symbols Use1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-259 of human USE1 (NP_0609371).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290627, 67023
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.