Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USE1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982123
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2123 Supplier Bioss Supplier No. BS12305R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MDS032 belongs to the USE1 family and is a component of a SNARE complex which may be involved in targeting and fusion of Golgi-derived retrograde transport vesicles with the ER.

Specifications

Antigen USE1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Use1
Gene Accession No. Q9NZ43
Gene Alias 2010315L10Rik; 5730403H22Rik; AV002165; BOS_7155; D12; Ed2; embryonic development factor 2; fj43b06; hypothetical protein MGC56402; MDS032; p31; Protein D12; protein p31; Protein transport protein USE1; putative MAPK activating protein PM26; Putative MAPK-activating protein PM26; Q-snare; RGD1306660; SLT1; SNAP receptor USE1; SNARE-like tail-anchored protein 1; SNARE-like tail-anchored protein 1 homolog; Unconventional SNARE in the endoplasmic reticulum protein 1; unconventional SNARE in the ER 1; unconventional SNARE in the ER 1 homolog; unconventional SNARE in the ER 1 homolog (S. cerevisiae); use1; Use1l; USE1-like protein; Vesicle transport protein USE1; wu:fj43b06; YGL098W; zgc:56402
Gene Symbols Use1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human USE1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55850
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.