Learn More
Description
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC23318 and based on accession number NM_18467. The protein contains a C-terminal DDK tag. |
| Quantity | 0.1 mg |
| Purity | Protein |
| Test Specificity | Homo sapiens unconventional SNARE in the ER 1 homolog (S. cerevisiae) (USE1), mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Protein State | Native |
| For Use With (Application) | Western Blot |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
