Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583340
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583340 Supplier Invitrogen™ Supplier No. PA583340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IQSAATFPDP NVKYVFRTEN GGQVMYRVIQ VSEGQLDGQT EGTGAISGYP ATQSMTQAVI QGAFTSDDAV DT Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 97%.

This gene encodes a member of the basic helix-loop-helix leucine zipper family, and can function as a cellular transcription factor. The encoded protein can activate transcription through pyrimidine-rich initiator (Inr) elements and E-box motifs. This gene has been linked to familial combined hyperlipidemia (FCHL). Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USF1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene USF1
Gene Accession No. P22415
Gene Alias bHLHb11; Class B basic helix-loop-helix protein 11; FCHL; FCHL1; HYPLIP1; Major late transcription factor 1; MLTF; MLTFI; UEF; Upstream stimulatory factor 1; upstream transcription factor 1; USF; USF1; usf1-bd
Gene Symbols USF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human USF1. Recombinant protein control fragment (Product #RP-104668).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7391
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.