Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USF2 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP192650AF488
Description
USF2 Polyclonal antibody specifically detects USF2 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
USF2 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
BHLHB12, bHLHb12upstream stimulatory factor 2, Class B basic helix-loop-helix protein 12, FIPc-fos interacting protein, FOS-interacting protein, Major late transcription factor 2, Upstream transcription factor 2, upstream transcription factor 2, c-fos interacting | |
The immunogen this antibody was made to, maps to a region between residue 296 and 346 of human Upstream Transcription Factor 2 using the numbering given in entry NP_003358.1 (GeneID 7392). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
7392 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction