Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ USF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565048
Description
Immunogen sequence: AFGDHNIQYQF RTETNGGQVT YRVVQVTDGQ LDGQGDTAGA VSVVSTAAFA GGQQAVTQVG VDG Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
USF2 consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.
Specifications
USF2 | |
Polyclonal | |
Unconjugated | |
USF2 | |
bHLHb12; c-fos interacting; c-fos interacting protein; class B basic helix-loop-helix protein 12; FIP; FOS-interacting protein; Major late transcription factor 2; transcription factor USF2; upstream stimulatory factor 2; Upstream transcription factor 2; upstream transcription factor 2, c-fos interacting; USF2; Usf-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7392 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15853 | |
USF2 | |
Recombinant Human USF2. Recombinant protein control fragment (Product #RP-104602). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction