Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ USF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565450
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565450 Supplier Invitrogen™ Supplier No. PA565450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GPAAPFPLAVI QNPFSNGGSP AAEAVSGEAR FAYFPASSVG DTTAVSVQTT DQSLQAGGQF YVMMTPQDVL QTGTQRTI Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

USF2 consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen USF2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene USF2
Gene Accession No. Q15853
Gene Alias bHLHb12; c-fos interacting; c-fos interacting protein; class B basic helix-loop-helix protein 12; FIP; FOS-interacting protein; Major late transcription factor 2; transcription factor USF2; upstream stimulatory factor 2; Upstream transcription factor 2; upstream transcription factor 2, c-fos interacting; USF2; Usf-2
Gene Symbols USF2
Host Species Rabbit
Immunogen Recombinant Human USF2. Recombinant protein control fragment (Product #RP-106128).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7392
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.